missing translation for 'onlineSavingsMsg'
Learn More

CD86 Mouse anti-Human, V450, Clone: 2331 (FUN-1), BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 560359

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15800139

  • 292.00 € / 1 Stück
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

2331 (FUN-1) recognizes a 75kDa cell surface protein, CD86 (B70/B7-2), expressed primarily on monocytes and activated B cells. Competitive binding assays demonstrate that, while both 2331 (FUN-1) and IT2.2 (anti-CD86, Mfr. No. 555663) recognize the same molecule, they react with different epitopes. CD86 is the second ligand for CD28 and CTLA-4 and may play an important role in co-stimulation of T cells in primary immune response. 2331 (FUN-1) blocks costimulation activity of CD86 in functional studies. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.

Host Species: Mouse
Clone: 2331(FUN-1)
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

Spezifikation

Spezifikation

CD86
Monoclonal
Violet Fluor 450 (V450)
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Human HBL-1 Cell Line
30 Tests
Immunology
Human
IgG1 κ
Flow Cytometry
2331 (FUN-1)
B7.2; B7-2; B-lymphocyte activation antigen B7-2; B70; BU63; CD28LG2; LAB72
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.