Learn More
CD94 Mouse anti-Human, Clone: 131412, R&D Systems™
Mouse Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB1058
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418295
  • 457.62 € / 500 Mikrogramm
Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
CD94 Monoclonal antibody MAB1058 Specifically detects CD94 in Human samples. It is validated for Blocking (Receptor-ligand), Flow Cytometry
Spezifikation
Spezifikation
CD94
131412
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD94, CD94 antigen, CD94natural killer cells antigen CD94, Killer cell lectin-like receptor subfamily D member 1, killer cell lectin-like receptor subfamily D, member 1, KLRD1, KP43, NK cell receptor
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Recognizes human CD94 both in its homodimeric form and as a heterodimer with either NKG2A or NKG2C.
Human
Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
LYOPH
Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready
Purified
KLRD1
BaF3 mouse pro-B cell line transfected with human CD94 and NKG2A
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3824
Reconstitute at 0.5 mg/mL in sterile PBS.