Learn More
CDCA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA519733
Weitere Details : CAS : 69-74-9 Netto-Gewicht : 0.01000kg
Beschreibung
PA5-19733 targets CDCA2 in WB applications and shows reactivity with Human samples. The PA5-19733 immunogen is synthetic peptide conjugated to KLH derived from within residues 950 to the C-terminus of Human CDCA2. PA5-19733 detects CDCA2 which has a predicted molecular weight of approximately 125 kDa.
Spezifikation
CDCA2 | |
Polyclonal | |
Unconjugated | |
Q69YH5 | |
CDCA2 | |
Synthetic peptide conjugated to KLH derived from within residues 950 to the C-terminus of Human CDCA2. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.2 mg/mL | |
CDCA2 | |
FLJ25804, MGC129906, MGC129907, Repo-Man, cell division cycle-associated protein 2; protein phosphatase 1 [gamma] (PPP1R3A) binding protein Repo-Man; recruits PP1 onto mitotic chromatin at anaphase protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
157313 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |