missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdk1 Mouse, Unlabeled, Clone: 1, BD
Mouse Monoclonal Antibody
330.00 € - 610.00 €
Spezifikation
Antigen | Cdk1 |
---|---|
Klon | 1 |
Konzentration | 250μg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15875219
|
BD Biosciences
610037 |
50 μg |
330.00 €
1 Set |
Verfügbar ab: 17-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15885219
|
BD Biosciences
610038 |
150 μg |
610.00 €
1 Set |
Verfügbar ab: 17-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Cdk1, also known as p34 [cdc2], is a ubiquitously expressed serine/threonine protein kinase. Cdk1/cdc2 has been identified as the catalytic subunit of the maturation-promoting factor (MPF), while cyclin B acts as the regulatory subunit. The binding of these two subunits is critical to the transition into M-phase of the mammalian cell cycle, and this factor's role is regulated by a series of phosphorylations and dephosphorylations. After binding to cyclin B, cdk1/cdc2 is phosphorylated on Thr-14, by Myt1, and Tyr-15, by wee1 or mik1, yielding an inactive pre-MPF complex. Phosphorylation of cdk1/cdc2 on Thr-161 is performed by a cdk7/cyclin H complex and is necessary for activation of the cdc2 complex. Dephosphorylation of Thr-14 and Tyr-15 by CDC25 occurs at the prophase/metaphase transition and completes activation of the cdc2/cyclin B complex, initiating the cell's entry into mitosis.Spezifikation
Cdk1 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
Cdc2, p34 |
1 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Human p34 [cdc2] aa. 283 to 297 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.