missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Chicken IgY (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 488, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen SA510070
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is cross-adsorbed and exhibits minimum reactivity to bovine, horse, human, mouse, pig, rabbit and rat.
Chicken IgY (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Chicken IgY (H+L) Cross-Adsorbed in Chicken samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Spezifikation
Chicken IgY (H+L) Cross-Adsorbed | |
Polyclonal | |
DyLight 488 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
Chicken | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4 | |
Chicken IgG (IgY)-heavy and light chain | |
500 μg | |
Secondary | |
4° C | |
IgG |