Learn More
Goat anti-Chicken IgM Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen PA184676
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Goat anti Chicken IgM antibody recognizes chicken immunoglobulin M and shows no cross reactivity with other chicken immunoglobulin classes in immunoelectrophoresis.Goat anti chicken IgM antibody may cross react with IgM from other species.
Chicken IgM Polyclonal antibody specifically detects Chicken IgM in Chicken samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot.Spezifikation
Chicken IgM | |
Polyclonal | |
HRP | |
Goat | |
Affinity chromatography | |
RUO | |
Chicken | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA and 0.05% ProClin 300 | |
Purified chicken IgM. | |
1 mg | |
Secondary | |
4° C, do not freeze | |
IgG |