Learn More
CHRM2 Mouse anti-Human, Clone: 585713, R&D Systems™
Mouse Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB6377
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13411346
  • 278.53 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 30-04-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
CHRM2 Monoclonal antibody MAB6377 Specifically detects CHRM2 in Human samples. It is validated for Immunohistochemistry
Spezifikation
Spezifikation
Muscarinic Acetylcholine Receptor M2/CHRM2
585713
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CHRM2
NS0 mouse myeloma cell line transfected with human CHRM2 Accession # P08172
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1129
Human
Immunohistochemistry
LYOPH
Immunohistochemistry
P08172
7TM receptor, cholinergic receptor, muscarinic 2, FLJ43243, HM2, mAChR M2, MGC120006, MGC120007, muscarinic acetylcholine receptor M2, muscarinic M2 receptor
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Sterile PBS to a final concentration of 0.5 mg/mL.