missing translation for 'onlineSavingsMsg'
Learn More

Complement C5a Monoclonal Antibody (2942), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA140161

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11529702

  • 582.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

MA1-40161 detects C5a/C5a des-Arg from human samples. MA1-40161 has been successfully used in Western blot, ELISA, immunohistochemistry (frozen), and functional assay applications. By Western blot, the antibody detects a band size of 14 kDa. The MA1-40161 immunogen is the Neo-epitope on C5a. Positive control: Eye tissue of patient with AMD; Negative control: Eye tissue of patient without AMD. Endotoxin <24 EU/mg. General comment: Please notice that under given conditions it is known that C5 can expose epitopes normally only found in the cleaved activation products.

Complement C5a Monoclonal antibody specifically detects Complement C5a in Human samples. It is validated for ELISA, Functional Assay, Immunohistochemistry (Frozen), Western Blot.
Spezifikation

Spezifikation

Complement C5a
Monoclonal
0.1 mg/mL
PBS with 0.1% BSA and no preservative
P01031
C5
Neo-epitope on C5a.
100 μg
Primary
Human
Antibody
IgG2a
ELISA, Functional Assay, Immunohistochemistry (Frozen), Western Blot
2942
Unconjugated
C5
anaphylatoxin; anaphylatoxin C5a analog; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4; C4B1; C4B12; C4B2; C4B3; C4B5; C4F; C5; C5a; C5a anaphylatoxin; C5b; C5D; CH; CO4; complement C5; Complement C5 alpha chain; Complement C5 alpha' chain; Complement C5 beta chain; complement component 5; CPAMD3; CPAMD4; DADB-112B14.11; ECLZB; FLJ17816; FLJ17822; FLJ60561; Hc; He; Hemolytic complement; MGC142298; MGC164979; prepro-C5; RGD1561905
Mouse
Protein G
RUO
727
4° C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote