missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX5B Monoclonal Antibody (16H12H9), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen 459110
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
Positive control: Isolated mitochondria from Human heart, Bovine heart, Rat heart, and Mouse heart
COX5B Monoclonal antibody specifically detects COX5B in Bovine, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western Blot.Spezifikation
COX5B | |
Monoclonal | |
1 mg/mL | |
HEPES buffered saline with 0.02% sodium azide | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgG2b κ |
Flow Cytometry, Western Blot | |
16H12H9 | |
Unconjugated | |
COX5B; COXVB; cytochrome c oxidase polypeptide Vb; cytochrome c oxidase polypeptide VB, mitochondrial; cytochrome c oxidase polypeptide VIa; cytochrome c oxidase precursor (EC 1.9.3.1); cytochrome c oxidase subunit 5B; cytochrome c oxidase subunit 5B, mitochondrial; cytochrome c oxidase subunit Vb; cytochrome c oxidase subunit VIA* | |
Bovine Complex IV (native holoenzyme protein, purified from heart) | |
100 μg | |
Primary | |
4° C, do not freeze | |
Liquid |