missing translation for 'onlineSavingsMsg'
Learn More

anti-DAX1/NR0B1, Clone: H7431, R&D Systems™

Marke:  RD Systems (Bio-Techne) PP-H7431-00

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412446

  • 517.42 € / 100 Mikrogramm


Voraussichtlicher Liefertermin 30-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Applications:

Direct ELISA, Western Blotting

Spezifikation

Spezifikation

DAX1/NR0B1
H7431
Western Blot
Purified
NR0B1
Recombinant human DAX1 (amino acids 1 - 80) and mouse myeloma cells (NS-1)
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
190
Western Blot
Unconjugated
A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide
AHC, AHCDSS, AHCH, AHX, DAX1DAX-1, dosage-sensitive sex reversal, DSS-AHC critical region on the X chromosome protein 1, GTD, HHG, NR0B1, NROB1, nuclear hormone receptor, Nuclear receptor DAX-1, nuclear receptor subfamily 0 group B member 1, nuclear receptor subfamily 0, group B, member 1
Mouse
IgG2a
100 ug
Store the unopened product at -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Monoclonal
Human
Videos