Learn More
DCC Mouse anti-Human, Clone: 531505, R&D Systems™
Mouse Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB5884
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13411126
  • 386.61 € / 100 Mikrogramm
Voraussichtlicher Liefertermin 29-10-2019
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
DCC Monoclonal antibody MAB5884 Specifically detects DCC in Human samples. It is validated for Immunohistochemistry
Spezifikation
Spezifikation
DCC
531505
Unconjugated
Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
DCC
E. coli-derived recombinant human DCC Ser1323-Phe1447 Accession # P43146
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1630
Human
Immunohistochemistry
LYOPH
Immunohistochemistry 8-25 ug/mL
P43146
Colorectal cancer suppressor, CRC18, CRCR1, deleted in colorectal cancer protein, deleted in colorectal carcinoma, IGDCC1colorectal tumor suppressor, Immunoglobulin superfamily DCC subclass member 1, immunoglobulin superfamily, DCC subclass, member 1, netrin receptor DCC, Tumor suppressor protein DCC
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening.
Monoclonal
Detects human DCC in direct ELISAs.