Learn More
OPRD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA127970
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-27970 detects delta Opioid receptor from rat and mouse samples. Expected to cross react with human samples due to 100% sequence homology. PA1-27970 has been successfully used in immunocytochemistry applications. The PA1-27970 immunogen is synthetic peptide: TACTPSDGPGGGAAA, corresponding to amino acids 358-372 of Rat Delta Opioid Receptor. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Spezifikation
OPRD1 | |
Polyclonal | |
OPRD1 | |
OPRD1 | |
synthetic peptide TACTPSDGPGGGAAA, corresponding to amino acids 358-372 of Rat Delta Opioid Receptor. | |
RUO | |
18386, 24613, 4985 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
P41143, P32300, P33533 | |
Rabbit | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |