Learn More
OPRD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA14560
Additional Details : Netto-Gewicht : 0.01000kg
Description
PA1-4560 detects delta Opioid Receptor from human, mouse, and rat samples. PA1-4560 has been successfully used in Western blotting applications. This antibody will detect a dominant band of ∽48 kDa representing the monomer form of DOR-1, and may also detect dimers or trimers as heterodimerization and homodimerization is common. The PA1-4560 immunogen is a synthetic peptide corresponding to residues T(361) P S D G P G G G A A A(372) of human DOR-1.
OPRD1 Polyclonal antibody specifically detects OPRD1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
OPRD1 | |
Polyclonal | |
Unconjugated | |
OPRD1 | |
delta 1 opioid receptor; delta opioid receptor 1; delta-type opioid receptor; DOR; D-OR-1; DOR-1; K56; mDOR; MSL-2; Nbor; Opioid receptor; opioid receptor A; opioid receptor delta 1; opioid receptor, delta 1; OPRD; Oprd1; Ror-a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18386, 24613, 4985 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P32300, P33533, P41143 | |
OPRD1 | |
Synthetic peptide corresponding to residues T(361) P S D G P G G G A A A(372) of human DOR-1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |