missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dehydrogenase/reductase (SDR family) member 9, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human DHRS9 protein.
Marke: Abnova H00010170-B01P.50ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Sequence: MLFWVLGLLILCGFLWTRKGKLKIEDITDKYIFITGCDSGFGNLAARTFDKKGFHVIAACLTESGSTALKAETSERLRTVLLDVTDPENVKRTAQWVKNQVGEKGLWGLINNAGVPGVLAPTDWLTLEDYREPIEVNLFGLISVTLNMLPLVKKAQGRVINVSSVGGRLAIVGGGYTPSKYAVEGFNDSLRRDMKAFGVHVSCIEPGLFKTNLADPVKVIEKKLAIWEQLSPDIKQQYGEGYIEKSLDKLKGNKSYVNMDLSPVVECMDHALTSLFPKTHYAAGKDAKIFWIPLSHMPAALQDFLLLKQKAELANPKAVSpezifikation
dehydrogenase/reductase (SDR family) member 9 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human DHRS9 protein. | |
DHRS9 | |
3ALPHA-HSD/RDH15/RDHL/RDHTBE/RETSDR8/SDR9C4 | |
Mouse | |
Affinity Purified | |
RUO | |
10170 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_005771.3 | |
DHRS9 | |
DHRS9 (NP_005762.2, 1 a.a. ∼ 319 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody |