missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Donkey IgG (H+L) Secondary Antibody, DyLight™ 594, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen SA510064
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Anti-Donkey secondary antibodies are affinity-purified antibodies with well-characterized specificity for equine (Donkey) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Spezifikation
Donkey IgG (H+L) | |
Polyclonal | |
DyLight 594 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Donkey | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4 | |
Donkey IgG-heavy and light chain | |
500 μg | |
Secondary | |
4° C | |
IgG |