missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Donkey IgG (H+L) Secondary Antibody, DyLight™ 680, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen SA510066
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Donkey IgG (H+L) Polyclonal antibody specifically detects Donkey IgG (H+L) in Donkey samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Spezifikation
Donkey IgG (H+L) | |
Polyclonal | |
DyLight 680 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Donkey | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4 | |
Donkey IgG-heavy and light chain | |
500 μg | |
Secondary | |
4° C | |
IgG |