Learn More
E. coli serotype O/K Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA127226
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-27226 detects E. coli from bacteria samples. It is reactive to all E.Coli species in the immunogen, plus 0157-H7 and K12 tested by immunoblot, reacts with more then 50 proteins by Western Blot
E. coli serotype O/K Polyclonal antibody specifically detects E. coli serotype O/K in Bacteria samples. It is validated for Immunocytochemistry.Spezifikation
E. coli serotype O/K | |
Polyclonal | |
Unconjugated | |
Escherichia coli | |
Heat killed sonicate blend of 6 different strains of E. coli: TOP 10F, XS127/P3, HB101, JM109, CJ236/P3, and ATCC 37197. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
4-5 mg/mL | |
0.01M PBS with 0.1% sodium azide; pH 7.2 | |
Goat | |
IgG fraction | |
RUO | |
Bacteria | |
Antibody | |
IgG |