missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EAR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534008
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
EAR2 Polyclonal antibody specifically detects EAR2 in Bovine, Canine, Hamster, Human, Mouse, Non-human Primate, Rat, Xenopus laevis, Zebrafish samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
EAR2 | |
Polyclonal | |
Unconjugated | |
NR2F6 | |
AV090102; avian erythroblastic leukemia viral (v-erb-a) oncogene homolog-like 2; COUP transcription factor 3; COUPg; COUP-TF3; EAR2; EAR-2; Erba related 2; ERBAL2; ERBA-related gene-2; Nr2f6; Nuclear receptor subfamily 2 group F member 6; nuclear receptor subfamily 2, group F, member 6; nuclear receptor V-erbA-related; orphan nuclear receptor EAR-2; Ovalbumin upstream promoter gamma nuclear receptor; ovalbumin upstream promoter gamma nuclear receptor rCOUPg; v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 2; V-erbA-related protein 2; V-erbA-related protein EAR-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100296331, 103163676, 13864, 2063, 245980, 609720 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O09017, P10588, P43136 | |
NR2F6 | |
Synthetic 16 amino acid peptide from ligand-binding domain of human NR2F6. | |
50 μg | |
Primary | |
Bovine, Canine, Hamster, Human, Mouse, Non-human Primate, Rat, X. laevis, Zebrafish | |
Antibody | |
IgG |