Learn More
ECHS1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530014
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, HeLa, HepG2, mouse brain. Predicted reactivity: Human (99%), Mouse (87%), Rat (85%), Pig (81%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ECHS1 Polyclonal antibody specifically detects ECHS1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
ECHS1 | |
Polyclonal | |
Unconjugated | |
Echs1 | |
C80529; ECHS1; ECHS1D; enoyl CoA hydratase, short chain, 1, mitochondrial; enoyl Coenzyme A hydratase, short chain 1; enoyl Coenzyme A hydratase, short chain, 1, mitochondrial; enoyl-CoA hydratase 1; Enoyl-CoA hydratase short chain 1 mitochondrial; enoyl-CoA hydratase, mitochondrial; enoyl-CoA hydratase, short chain 1; Enoyl-CoA hydratase, short chain 1, mitochondrial; enoyl-CoA hydratase, short chain, 1, mitochondrial; mitochondrial short-chain enoyl-coenzyme A hydratase 1; SCEH; short chain enoyl-CoA hydratase; short-chain enoyl-CoA hydratase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1892, 93747 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1.01 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P30084, Q8BH95 | |
Echs1 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 157 of Human ECHS1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |