Learn More
ECM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA542882
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: QDRSQGGWGH RLDGFPPGRP SPDNLNQICL PNRQHVVYGP WNLPQSSYSH Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 93%; Rat: 100%
ECM1 Polyclonal antibody specifically detects ECM1 in Human samples. It is validated for Western Blot.Spezifikation
ECM1 | |
Polyclonal | |
Unconjugated | |
ECM1 | |
AI663821; ECM1; Extracellular matrix protein 1; LOW QUALITY PROTEIN: extracellular matrix protein 1; extracellular matrix protein 1; p85; RP11-54A4.6; Secretory component p85; testicular tissue protein Li 61; URBWD | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1893 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q16610 | |
ECM1 | |
Synthetic peptide directed towards the middle region of human ECM1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |