missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDF1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA537908
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
This antibody is predicted to react with mouse, rat and pig based on sequence homology.
EDF1 Polyclonal antibody specifically detects EDF1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
EDF1 | |
Polyclonal | |
Unconjugated | |
EDF1 | |
0610008L11Rik; AA409425; calmodulin-associated peptide 19; calmodulin-associated peptide-19; CAP-19; CFAP280; Edf1; EDF-1; endothelial differentiation related factor 1; endothelial differentiation-related factor 1; Endothelial differentiation-related factor 1 homolog; hypothetical protein 1-9; hypothetical protein MGC73192; MBF1; MGC9058; multiprotein bridging factor 1; multiprotein-bridging factor 1; zgc:73192; zgc:86829 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
8721 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O60869 | |
EDF1 | |
Peptide with sequence C-NKQHSITKNTAKLDR, from the internal region of EDF1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |