Learn More
EEA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529013
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HepG2, A375, mouse brain, PC-12, Rat-2, A549, MNT-1. Predicted reactivity: Mouse (100%), Rat (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EEA1 Polyclonal antibody specifically detects EEA1 in Hamster, Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
EEA1 | |
Polyclonal | |
Unconjugated | |
EEA1 | |
A430109M19Rik; B230358H09Rik; Early endosome antigen 1; early endosome antigen 1, 162kD; early endosome-associated protein; EEA1; Endosome-associated protein p162; MST105; MSTP105; ZFYVE2; Zinc finger FYVE domain-containing protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
216238, 314764, 8411 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.0 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q15075, Q8BL66 | |
EEA1 | |
Synthetic peptide corresponding to a region within amino acids 1342 and 1411 of Human EEA1 | |
100 μL | |
Primary | |
Hamster, Human, Mouse, Rat | |
Antibody | |
IgG |