missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGR3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538661
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
This gene encodes a transcriptional regulator that belongs to the EGR family of C2H2-type zinc-finger proteins. It is an immediate-early growth response gene which is induced by mitogenic stimulation. The protein encoded by this gene participates in the transcriptional regulation of genes in controling biological rhythm. It may also play a role in a wide variety of processes including muscle development, lymphocyte development, endothelial cell growth and migration, and neuronal development. Alternative splicing results in multiple transcript variants encoding distinct isoforms.[provided by RefSeq, Dec 2010].Spezifikation
EGR3 | |
Polyclonal | |
Unconjugated | |
EGR3 | |
Egr-3, PILOT | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13655, 1960, 25148 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q06889, P43300, P43301 | |
EGR3 | |
A synthetic peptide derived from the internal region of human EGR3 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |