Learn More
EIF2S1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529593
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (94%), Chicken (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
EIF2S1 Polyclonal antibody specifically detects EIF2S1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
EIF2S1 | |
Polyclonal | |
Unconjugated | |
EIF2S1 | |
0910001O23Rik; 2410026C18Rik; 35kDa; EIF2; EIF-2; eIF2 alpha; eIF2 alpha subunit; EIF2A; eIF-2A; eIF2alpha; EIF-2alpha; eIF-2-alpha; EIF2S1; eif2s1b; Eukaryotic translation initiation factor 2 subunit 1; eukaryotic translation initiation factor 2 subunit 1b; eukaryotic translation initiation factor 2 subunit alpha; eukaryotic translation initiation factor 2, subunit 1 alpha; eukaryotic translation initiation factor 2, subunit 1 alpha b; eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 2A; fb19d01; translation initiation factor subunit; wu:fb19d01 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1965 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.62 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P05198 | |
EIF2S1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 183 of Human EIF2 alpha | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |