missing translation for 'onlineSavingsMsg'
Learn More

eIF2 gamma Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA531177

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13405697

  • 512.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: HeLa, HepG2, HCT116. Predicted reactivity: Rat (100%), Zebrafish (98%), Xenopus laevis (97%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

eIF2 gamma Polyclonal antibody specifically detects eIF2 gamma in Human samples. It is validated for Western Blot.
Spezifikation

Spezifikation

eIF2 gamma
Polyclonal
Unconjugated
EIF2S3
EIF2; EIF2G; eIF-2gA; EIF2gamma; eIF-2-gamma; eIF-2-gamma X; eIF-2gX; Eif2s3; Eif2s3x; Eukaryotic translation initiation factor 2 subunit 3; Eukaryotic translation initiation factor 2 subunit 3, X-linked; Eukaryotic translation initiation factor 2 subunit gamma; eukaryotic translation initiation factor 2 subunit gamma X; Eukaryotic translation initiation factor 2 subunit gamma, X-linked; eukaryotic translation initiation factor 2, subunit 3 gamma, 52kDa; eukaryotic translation initiation factor 2, subunit 3, structural gene X-linked; eukaryotic translation initiation factor 2G; PP42
Rabbit
Antigen affinity chromatography
RUO
1968
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
P41091
EIF2S3
Recombinant fragment corresponding to a region within amino acids 1 and 307 of Human EIF2S3
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote