missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF3a Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517212
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
eIF3a Polyclonal antibody specifically detects eIF3a in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Spezifikation
eIF3a | |
Polyclonal | |
Unconjugated | |
EIF3A | |
150/170; A830012B05Rik; Centrosomin; centrosomin homolog; Csma; cytoplasmic protein p167; Eif3; eIF3 p167; eIF3 p180; eIF3 p185; EIF3, p180 subunit; eIF3a; eIF3a {ECO:0000255; eIF3-p170; EIF3S10; eIF-3-theta; eIF3-theta; eIF-3-theta {ECO:0000255; eukaryotic translation initiation factor 3 subunit 10; EUKARYOTIC TRANSLATION INITIATION FACTOR 3 SUBUNIT 10 (EIF-3 THETA) (EIF3 P167) (EIF3 P180) (EIF3 P185) (P162 PROTEIN) (CENTROSOMIN); eukaryotic translation initiation factor 3 subunit 10 {ECO:0000255; eukaryotic translation initiation factor 3 subunit A; eukaryotic translation initiation factor 3 subunit A {ECO:0000255; eukaryotic translation initiation factor 3, subunit 10 (theta); eukaryotic translation initiation factor 3, subunit 10 (theta, 150/170kD); eukaryotic translation initiation factor 3, subunit 10 (theta, 170kD); eukaryotic translation initiation factor 3, subunit 10 theta, 150/170kDa; eukaryotic translation initiation factor 3, subunit 10, 170kD; eukaryotic translation initi | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13669, 292148, 8661 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
15.2 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P23116, Q14152, Q1JU68 | |
EIF3A | |
Synthetic peptide corresponding to a sequence of human eIF3A | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |