missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF3d Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA523293
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
eIF3d Polyclonal antibody specifically detects eIF3d in Human, Mouse, Non-human Primate, Rat, Zebrafish samples. It is validated for Western Blot.Spezifikation
eIF3d | |
Polyclonal | |
Unconjugated | |
EIF3D | |
66/67kDa; AA407891; actinin alpha 4; ACTININ-4; Actn4; alpha actinin 4; alpha-actinin-4; eIF3 p66; EIF3D; eIF3d {ECO:0000255; eIF3p66; eIF3-p66; Eif3s7; eIF-3-zeta; eIF3-zeta; eIF-3-zeta {ECO:0000255; eukaryotic translation initiation factor 3 subunit 7; eukaryotic translation initiation factor 3 subunit 7 {ECO:0000255; Eukaryotic translation initiation factor 3 subunit D; eukaryotic translation initiation factor 3 subunit D {ECO:0000255; eukaryotic translation initiation factor 3, subunit 7 (zeta); eukaryotic translation initiation factor 3, subunit 7 zeta; eukaryotic translation initiation factor 3, subunit 7 zeta, 66/67kDa; eukaryotic translation initiation factor 3, subunit D; F-actin cross-linking protein; focal segmental glomerulosclerosis 1; FSGS; FSGS1; HAMAP-Rule:MF_03003}; non-muscle alpha-actinin 4; RP5-1119A7.12-003; si:dkey-165I8.6; translation initiation factor eIF3 p66; translation initiation factor eIF3 p66 subunit; wu:fb12a03; zgc:64097 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
336498, 362952, 55944, 8664 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
O15371, O70194, Q6AYK8, Q6TH15 | |
EIF3D | |
A portion of amino acids 500-548 of human eIF3D | |
100 μg | |
Primary | |
Human, Mouse, Non-human Primate, Rat, Zebrafish | |
Antibody | |
IgG |