missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF3e Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA519411
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, canine and porcine based on sequence homology.
eIF3e Polyclonal antibody specifically detects eIF3e in Human samples. It is validated for Western Blot.Spezifikation
eIF3e | |
Polyclonal | |
Unconjugated | |
EIF3E | |
48kDa; eIF-3 p48; eIF3e; eIF3-p46; EIF3-P48; EIF3S6; eukaryotic translation initiation factor 3 subunit 6; Eukaryotic translation initiation factor 3 subunit E; eukaryotic translation initiation factor 3 subunit E isoform 2 transcript; eukaryotic translation initiation factor 3 subunit E-like protein; eukaryotic translation initiation factor 3, subunit 6 (48kD); eukaryotic translation initiation factor 3, subunit 6 48kDa; eukaryotic translation initiation factor 3, subunit E; eukaryotic translation initiation factor subunit E-like protein; Int6; mammary tumor integration site 6; mammary tumor-associated protein INT6; mammary tumor-associated protein INT-6; MMTV integration site 6; murine mammary tumor integration site 6 (oncogene homolog); NEWGENE_1311521; Viral integration site protein INT-6; viral integration site protein INT-6 homolog | |
Goat | |
Antigen affinity chromatography | |
RUO | |
3646 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P60228 | |
EIF3E | |
Synthetic peptide sequence (KLNQNSRSEAPN) corresponding to the C-terminus amino acids of EIF3E | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |