Learn More
eIF3j Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke:  Invitrogen PA517062
Weitere Details : Netto-Gewicht : 0.01000kg
Artikelnummer. 11551531
  • 473.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
It is not recommended to aliquot this antibody.
eIF3j Polyclonal antibody specifically detects eIF3j in Human, Mouse, Rat samples. It is validated for Immunoprecipitation, Western Blot.
Spezifikation
Spezifikation
eIF3j
Polyclonal
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
A0JPM9, O75822, Q3UGC7, Q66JS6
EIF3J, Eif3j1, Eif3j2
Synthetic peptide corresponding to the sequence of human eIF3J
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Immunoprecipitation, Western Blot
Unconjugated
EIF3J
1810016I04Rik; 2700079K05Rik; AA409117; AA409446; C78575; eIF3 p35; eIF3 p35 {ECO:0000255; eIF-3-alpha; eIF3-alpha; eIF-3-alpha {ECO:0000255; eIF-3-alpha-A; eIF-3-alpha-B; Eif3j; eIF3j {ECO:0000255; Eif3j1; Eif3j-1; Eif3j2; Eif3j-2; eIF3j-A; eIF3j-B; eIF3-p35; EIF3S1; Eif3s1-1; Eif3s1-2; ENSMUSG00000043424; Eukaryotic translation initiation factor 3 subunit 1; eukaryotic translation initiation factor 3 subunit 1 {ECO:0000255; eukaryotic translation initiation factor 3 subunit 1-A; Eukaryotic translation initiation factor 3 subunit 1-B; eukaryotic translation initiation factor 3 subunit J; eukaryotic translation initiation factor 3 subunit J {ECO:0000255; Eukaryotic translation initiation factor 3 subunit J-A; Eukaryotic translation initiation factor 3 subunit J-B; eukaryotic translation initiation factor 3, subunit 1 (alpha, 35kD); eukaryotic translation initiation factor 3, subunit 1 alpha; eukaryotic translation initiation factor 3, subunit 1 alpha, 35kDa; eukaryotic translation init
Rabbit
Antigen affinity chromatography
RUO
100042807, 691947, 78655, 8669
-20°C