Learn More
eIF4B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529540
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: NT2D1, IMR32, MCF-7, U87-MG, SK-N-SH, SK-N-AS, 293T, A431, HeLa, HepG2, human EIF4B-transfected 293T. Predicted reactivity: Mouse (97%), Rat (97%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
eIF4B Polyclonal antibody specifically detects eIF4B in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
eIF4B | |
Polyclonal | |
Unconjugated | |
Eif4b | |
2310046H11Rik; AL024095; C85189; Eif4a2; EIF4B; eIF-4B; Eukaryotic translation initiation factor 4B; hCG_2014609; hypothetical protein; PRO1843 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1975, 75705 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.16 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P23588, Q8BGD9 | |
Eif4b | |
Recombinant fragment corresponding to a region within amino acids 1 and 198 of Human EIF4B | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |