missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA519666
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with mouse, rat, cow and zebrafish based on sequence homology.
eIF4B Polyclonal antibody specifically detects eIF4B in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
eIF4B | |
Polyclonal | |
Unconjugated | |
Eif4b | |
2310046H11Rik; AL024095; C85189; Eif4a2; EIF4B; eIF-4B; Eukaryotic translation initiation factor 4B; hCG_2014609; hypothetical protein; PRO1843 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1975 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
P23588 | |
Eif4b | |
Synthetic peptide conjugated to KLH derived from within residues 300 - 400 of Human eIF4B. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |