missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA519772
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with mouse, rat and cow based on sequence homology.
Spezifikation
eIF4B | |
Polyclonal | |
Unconjugated | |
P23588 | |
EIF4B | |
Synthetic peptide conjugated to KLH derived from within residues 550 to the C-terminus of Human eIF4B. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
0.1 mg/mL | |
EIF4B | |
EIF-4B, PRO1843 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1975 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |