missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4EBP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA511830
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine based on sequence homology.
4EBP1 Polyclonal antibody specifically detects 4EBP1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
4EBP1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q13541 | |
EIF4EBP1 | |
KLH conjugated synthetic peptide between 48-77 amino acids from human EIF4EBP1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
EIF4EBP1 | |
4EBP1; 4E-BP1; AA959816; BP-1; eIF4E-binding protein 1; EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1; eukaryotic translation initiation factor 4E-binding protein 1; MGC431; MGC4316; P/OKCL.6; PHAS-1; PHAS-I; Phosphorylated heat- and acid-stable protein regulated by insulin 1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
1978 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |