Learn More
eIF5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530223
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: U87-MG, mouse brain, rat brain. Predicted reactivity: Mouse (97%), Rat (97%), Chicken (92%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
eIF5 Polyclonal antibody specifically detects eIF5 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
eIF5 | |
Polyclonal | |
Unconjugated | |
Eif5 | |
2810011H21Rik; D12Ertd549e; EIF5; EIF-5; EIF-5A; eukaryotic initiation factor 5 (eIF-5); eukaryotic translation initiation factor 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
108348073, 1983, 217869, 56783 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P55010, P59325, Q07205 | |
Eif5 | |
Recombinant protein fragment corresponding to a region within amino acids 184 and 415 of Human EIF5 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |