missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF5A Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA519191
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
eIF5A Polyclonal antibody specifically detects eIF5A in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
eIF5A | |
Polyclonal | |
Unconjugated | |
EIF5A | |
AA410058; D19Wsu54e; Eif4d; eIF-4D; Eif5a; eIF-5A; EIF5A1; eIF-5A1; eIF-5A-1; eIF5AI; eukaryotic initiation factor 5A; Eukaryotic initiation factor 5A isoform 1; eukaryotic translation initiation factor 5A; eukaryotic translation initiation factor 5A-1; MGC104255; MGC99547; OTTHUMP00000128384; Rev-binding factor; uORF A | |
Goat | |
Antigen affinity chromatography | |
RUO | |
1984 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P63241 | |
EIF5A | |
Synthetic peptide sequence (RPPHRASFLKRLESK) corresponding to the internal amino acids of EIF5A | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |