missing translation for 'onlineSavingsMsg'
Learn More

eIF5A2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA522118

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13279578

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

PA5-22118 targets EIF5A2 in ICC, IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-22118 immunogen is a synthetic peptide corresponding to a region within amino acids 89 and 153 of EIF5A2 (Uniprot IDNo.Q9GZV4). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Spezifikation

Spezifikation

EIF5A2
Polyclonal
Unconjugated
Q9GZV4
EIF5A2
Synthetic peptide corresponding to a region within amino acids 89 and 153 of EIF5A2 (Uniprot IDNo.Q9GZV4)
100 μL
Primary
Human
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.87 mg/mL
EIF5A2
EIF-5A2, eIF5AII, eIF-5A-2, eukaryotic initiation factor 5A, eukaryotic translation initiation factor 5A-2
Rabbit
Antigen affinity chromatography
RUO
56648
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote