missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF5B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA536456
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
This antibody detects endogenous protein at a molecular weight of 139 kDa. Purity is >95% by SDS-PAGE.
eIF5B Polyclonal antibody specifically detects eIF5B in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
eIF5B | |
Polyclonal | |
Unconjugated | |
EIF5B | |
A030003E17Rik; Annexin V-binding protein ABP-7; BC018347; DKFZp434I036; eIF 5B; Eif5b; eIF-5B; Eukaryotic translation initiation factor 5B; FLJ10524; If2; KIAA0741; translation initiation factor IF2; Translation initiation factor IF-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
226982, 308306, 9669 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
B2GUV7, O60841, Q05D44 | |
EIF5B | |
Synthetic peptide corresponding to amino acids 1061-1110 of human eIF5B | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |