missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA537371
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
This antibody detects endogenous protein at a molecular weight of 27 kDa. Purity is >95% by SDS-PAGE.
eIF6 Polyclonal antibody specifically detects eIF6 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
eIF6 | |
Polyclonal | |
Unconjugated | |
Eif6 | |
1110004P11; AA408895; b(2)gcn; B(2)GCN homolog; B4 integrin interactor; CAB; EIF3A; Eif6; eIF-6; eIF-6 {ECO:0000255; eukaryotic translation initiation factor 3A; eukaryotic translation initiation factor 6; eukaryotic translation initiation factor 6 {ECO:0000255; HAMAP-Rule:MF_03132}; imc-415; integrin beta 4 binding protein; Itgb4bp; OK/SW-cl.27; p27 beta-4 integrin-binding protein; p27(BBP); p27BBP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3692 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
P56537 | |
Eif6 | |
Recombinant full length human eIF6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |