Learn More
Neutrophil elastase Monoclonal Antibody (265-3K1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA140220
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-40220 detects Elastase from human samples. MA1-40220 has been successfully used in ELISA, Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry and Western blot applications. The MA1-40220 immunogen is human leukocyte elastase.
Neutrophil elastase Monoclonal antibody specifically detects Neutrophil elastase in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Neutrophil elastase | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% BSA and 0.02% sodium azide | |
P08246 | |
ELANE | |
Human leukocyte elastase. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
265-3K1 | |
Unconjugated | |
ELANE | |
bone marrow serine protease; ELA2; ELA2A; ELANE; Elastase; elastase 2, neutrophil; elastase, neutrophil expressed; Elastase2; elastase-2; F430011M15Rik; GE; granulocyte-derived elastase; HLE; HNE; Human leukocyte elastase; Leukocyte elastase; medullasin; NE; neutrophil elastase; PE-1; PMN elastase; PMN-E; polymorphonuclear elastase; SCN1; serine protease | |
Mouse | |
Protein G | |
RUO | |
1991 | |
4° C | |
Liquid |