missing translation for 'onlineSavingsMsg'
Learn More

Neutrophil elastase Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA529659

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13433877

  • 514.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Recommended positive controls: A549, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Neutrophil elastase Polyclonal antibody specifically detects Neutrophil elastase in Human samples. It is validated for Immunoprecipitation, Western Blot.
Spezifikation

Spezifikation

Neutrophil elastase
Polyclonal
Unconjugated
ELANE
bone marrow serine protease; ELA2; ELA2A; ELANE; Elastase; elastase 2, neutrophil; elastase, neutrophil expressed; Elastase2; elastase-2; F430011M15Rik; GE; granulocyte-derived elastase; HLE; HNE; Human leukocyte elastase; Leukocyte elastase; medullasin; NE; neutrophil elastase; PE-1; PMN elastase; PMN-E; polymorphonuclear elastase; SCN1; serine protease
Rabbit
Antigen affinity chromatography
RUO
1991
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunoprecipitation, Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
P08246
ELANE
Recombinant fragment corresponding to a region within amino acids 61 and 267 of Human Elastase
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote