missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELF3 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518065
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
ELF3 Polyclonal antibody specifically detects ELF3 in Human samples. It is validated for Western Blot.Spezifikation
ELF3 | |
Polyclonal | |
Unconjugated | |
ELF3 | |
E74 like ETS transcription factor 3; E74-like factor 3; E74-like factor 3 (ets domain transcription factor); E74-like factor 3 (ets domain transcription factor, epithelial-specific ); E74-like factor 3 (ETS domain transcription factor, serine box, epithelial-specific); ELF3; elf3 {ECO:0000312; EMBL:AAH97980.1}; Epithelial-restricted with serine box; epithelium-restricted Ets protein ESX; Epithelium-specific Ets transcription factor 1; EPR-1; Ert; ESE-1; ESX; ets domain transcription factor, serine box (epithelial-specific); ETS-related transcription factor Elf-3; JEN; RP11-510N19.1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
1999 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P78545 | |
ELF3 | |
Synthetic peptide sequence (SGWKEEEVLQSRN) corresponding to the C-terminus amino acids of ELF3 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |