missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-ELK3 (Ser357) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538489
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
The protein encoded by this gene is a member of the ETS-domain transcription factor family and the ternary complex factor (TCF) subfamily. Proteins in this subfamily regulate transcription when recruited by serum response factor to bind to serum response elements. This protein is activated by signal-induced phosphorylation; studies in rodents suggest that it is a transcriptional inhibitor in the absence of Ras, but activates transcription when Ras is present.Spezifikation
ELK3 | |
Polyclonal | |
Unconjugated | |
ELK3 | |
ELK3, NET, SAP2, ETS domain-containing protein Elk-3, ETS-related protein ERP, ETS-related protein NET, Serum response factor accessory protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2004 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P41970 | |
ELK3 | |
A synthetic phosphopeptide derived from human ETS Domain Protein Elk-3 around the phosphorylation site of Ser357 (S-L-SP-P-V) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |