missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELK3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA535173
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
ELK3 Polyclonal antibody specifically detects ELK3 in Human samples. It is validated for Western Blot.Spezifikation
ELK3 | |
Polyclonal | |
Unconjugated | |
ELK3 | |
D430049E23Rik; ELK3; ELK3, ETS transcription factor; ELK3, ETS-domain protein; ELK3, ETS-domain protein (SRF accessory protein 2); ELK3, member of ETS oncogene family; Erp; Etrp; ETS domain-containing protein Elk-3; ETS transcription factor ELK3; ETS-domain protein ELK-3; ETS-related protein ERP; ETS-related protein NET; Net; SAP2; Sap-2; Serum response factor accessory protein 2; si:ch211-172b12.2; si:dkey-201e17.1; SRF accessory protein 2; zgc:198153; zgc:198183 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
2004 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide | |
P41970 | |
ELK3 | |
KLH conjugated synthetic peptide between 98-126 amino acids from the N-terminal region of human ELK3 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |