Learn More
ELL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA541530
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: PGSKEYQNVH EEVLQEYQKI KQSSPNYHEE KYRCEYLHNK LAHIKRLIGE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%
ELL2 Polyclonal antibody specifically detects ELL2 in Human samples. It is validated for Western Blot.Spezifikation
ELL2 | |
Polyclonal | |
Unconjugated | |
ELL2 | |
cb244; eleven-nineteen lysine-rich leukemia gene 2; ELL2; ELL-related RNA polymerase II, elongation factor; elongation factor for RNA polymerase II 2; elongation factor RNA for polymerase II 2; elongation factor RNA polymerase II 2; elongation factor, RNA polymerase II 2; elongation factor, RNA polymerase II, 2; fb78f03; im:3814960; RNA polymerase II elongation factor ELL2; RNA polymerase II elongation factor ELL2-like protein; sb:cb244; wu:fb78f03; wu:fb78f03 protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
22936 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O00472 | |
ELL2 | |
Synthetic peptide directed towards the C-terminal of human ELL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |