missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELMO2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518002
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology.
ELMO2 Polyclonal antibody specifically detects ELMO2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ELMO2 | |
Polyclonal | |
Unconjugated | |
ELMO2 | |
1190002F24Rik; CED12; CED-12; ced-12 homolog 2; CED-12 homolog A; CED12A; Ced-12A; Elmo2; ELMO-2; engulfment and cell motility 2; engulfment and cell motility 2, ced-12 homolog; engulfment and cell motility protein 2; FLJ11656; hCed-12A; KIAA1834; PH domain protein CED12A; protein ced-12 homolog A; RP11-394O2.2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
140579, 362271, 63916 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8BHL5, Q96JJ3 | |
ELMO2 | |
Synthetic peptide sequence (PKEPSSYDFVYHYG) corresponding to the C-terminus amino acids of ELMO2 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |