missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELMO3 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA517999
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with canine based on sequence homology.
ELMO3 Polyclonal antibody specifically detects ELMO3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ELMO3 | |
Polyclonal | |
Unconjugated | |
ELMO3 | |
9930107J06; BC058752; Ced12; CED-12; ced-12 homolog 3; Elmo3; ELMO-3; engulfment and cell motility 3; engulfment and cell motility 3, ced-12 homolog; engulfment and cell motility protein 3; FLJ13824 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
79767 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q96BJ8 | |
ELMO3 | |
Synthetic peptide sequence (TNFNFCYDCSIAEP) corresponding to the C-terminus amino acids of ELMO3 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |