missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELOVL4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA112913
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control for this product is brain tissue lysate.
ELOVL4 Polyclonal antibody specifically detects ELOVL4 in Human samples. It is validated for Western Blot.Spezifikation
ELOVL4 | |
Polyclonal | |
PBS with 0.08% sodium azide | |
Q9GZR5 | |
ELOVL4 | |
Synthetic peptide corresponding to the human ELOV4 protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ELOVL4 | |
3-keto acyl-CoA synthase Elovl4; ADMD; cancer/testis antigen 118; CT118; elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast)-like 4; elongation of very long chain fatty acids protein 4; elongation of very long chain fatty acids-like 4; ELOVL FA elongase 4; ELOVL fatty acid elongase 4; Elovl4; FLJ17667; FLJ92876; ISQMR; SCA34; STGD2; STGD3; Very long chain 3-ketoacyl-CoA synthase 4; very long chain 3-oxoacyl-CoA synthase 4; very-long-chain 3-oxoacyl-CoA synthase 4 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
6785 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |