missing translation for 'onlineSavingsMsg'
Learn More

ELOVL5 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA532741

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13494409

  • 473.00 € / 50 Mikrogramm
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey, Marmoset, Turkey, Chicken (100%) Elephant, Catfish (93%).

ELOVL5 Polyclonal antibody specifically detects ELOVL5 in Chicken, Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).
Spezifikation

Spezifikation

ELOVL5
Polyclonal
Unconjugated
ELOVL5
1110059L23Rik; 3-keto acyl-CoA synthase ELOVL5; AI747313; AU043003; dJ483K16.1; elongation of long chain fatty acids family member 5; elongation of very long chain fatty acids protein 5; elongation of very long chain fatty acids-like 5; ELOVL FA elongase 5; ELOVL family member 5, elongation of long chain fatty acids; ELOVL family member 5, elongation of long chain fatty acids (FEN1/Elo2, SUR4/Elo3-like, yeast); ELOVL family member 5, elongation of long chain fatty acids (yeast); ELOVL fatty acid elongase 5; ELOVL2; ELOVL5; Fatty acid elongase 1; hELO1; homolog of yeast long chain polyunsaturated fatty acid elongation enzyme; homolog of yeast long chain polyunsaturated fatty acid elongation enzyme 2; PRO0530; rELO1; SCA38; spinocerebellar ataxia 38; very long chain 3-ketoacyl-CoA synthase 5; Very long chain 3-oxoacyl-CoA synthase 5; very-long-chain 3-oxoacyl-CoA synthase 5
Rabbit
Antigen affinity chromatography
RUO
428646, 60481
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
Q9NYP7
ELOVL5
Synthetic 15 amino acid peptide from N-terminus of human ELOVL5.
50 μg
Primary
Chicken, Human, Non-human Primate
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote