Learn More
ELTD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532743
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset (100%); Rabbit (89%); Dog, Elephant, Panda, Horse (84%). For IHC(P), use proteinase K antigen retrieval. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
ELTD1 Polyclonal antibody specifically detects ELTD1 in Human, Non-human Primate samples. It is validated for ELISA, Immunohistochemistry (Paraffin).Spezifikation
ELTD1 | |
Polyclonal | |
Unconjugated | |
ADGRL4 | |
1110033N21Rik; ADGRL4; Adhesion G protein-coupled receptor L4; EGF, latrophilin and seven transmembrane domain containing 1; EGF, latrophilin and seven transmembrane domain-containing protein 1; EGF, latrophilin seven transmembrane domain containing 1; EGF, latrophilin seven transmembrane domain-containing protein 1; EGF,latrophilin and seven transmembrane domain-containing protein 1; EGF-TM7 receptor; EGF-TM7-latrophilin-related protein; Eltd1; ETL; ETL protein; ETL1; hCG_14667; KPG_003; UNQ202/PRO228 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64123 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9HBW9 | |
ADGRL4 | |
Synthetic 19 amino acid peptide from N-terminal extracellular domain of human ELTD1. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |