missing translation for 'onlineSavingsMsg'
Learn More

Embigin Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA532744

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13404419

  • 470.00 € / 50 Mikrogramm
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Monkey (100%); Orangutan (94%); Gibbon, Marmoset, Bat (88%); Bovine, Dog (82%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

Embigin Polyclonal antibody specifically detects Embigin in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).
Spezifikation

Spezifikation

Embigin
Polyclonal
Unconjugated
EMB
AL022799; EMB; Embigin; embigin homolog; glycoprotein 70; Gp70; Teratocarcinoma glycoprotein Gp-70
Rabbit
Antigen affinity chromatography
RUO
133418
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
Q6PCB8
EMB
Synthetic 17 amino acid peptide from C-Terminus of human EMB.
50 μg
Primary
Human, Non-human Primate
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote